Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sudan IV TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
67-66-3,85-83-6
CHCl3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
ClC(Cl)Cl
67-66-3,85-83-6
CHCl3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
ClC(Cl)Cl

S-251, 85-83-6

Spectrum™ Sudan IV TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS Max % 0.001,0.999
UN Number UN1888
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 1 L