Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUFU Monoclonal Antibody (C.60.7)
Rabbit Monoclonal Antibody
Supplier: Invitrogen™ MA514956
Description
It is not recommended to aliquot this antibody.
The Hedgehog signaling pathway plays an important role in early human development. The pathway is a signaling cascade that plays a role in pattern formation and cellular proliferation during development. This gene encodes a negative regulator of the hedgehog signaling pathway. Defects in this gene are a cause of medulloblastoma. Alternative splicing results in multiple transcript variants.
Specifications
SUFU | |
Monoclonal | |
Unconjugated | |
SUFU | |
OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; PRO1280; RP11-47A8.1; Su(fu); Sufu; SUFU negative regulator of hedgehog signaling; SUFUH; SUFUXL; suppressor of fused homolog; suppressor of fused homolog (Drosophila); UNQ650/PRO1280 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
24069, 361769, 51684 | |
-20°C | |
Liquid |
Western Blot | |
C.60.7 | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
Q9UMX1, Q9Z0P7 | |
SUFU | |
Synthetic peptide corresponding to residues surrounding Leu458 of human SUFU. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction