Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sulfacetamide, 98%

Click to view available options
Quantity:
50g
250g
144-80-9
C8H10N2O3S
214.239
MFCD00066501
SKIVFJLNDNKQPD-UHFFFAOYSA-N
sulfacetamide,sulphacetamide,acetosulfamine,n-acetylsulfanilamide,acetosulfamin,albucid,n-sulfanilylacetamide,n-4-aminophenyl sulfonyl acetamide,sulfacetimide,acetocid
5320
CHEBI:63845
N-(4-aminophenyl)sulfonylacetamide
CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N
144-80-9
C8H10N2O3S
214.239
MFCD00066501
SKIVFJLNDNKQPD-UHFFFAOYSA-N
sulfacetamide,sulphacetamide,acetosulfamine,n-acetylsulfanilamide,acetosulfamin,albucid,n-sulfanilylacetamide,n-4-aminophenyl sulfonyl acetamide,sulfacetimide,acetocid
5320
CHEBI:63845
N-(4-aminophenyl)sulfonylacetamide
CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N
Melting Point 182°C to 184°C
Quantity 250g
Beilstein 981718
Merck Index 14,8899
Formula Weight 214.24
Percent Purity 98%
Chemical Name or Material Sulfacetamide

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P281-P308+P313-P501c

H341

EINECSNumber : 205-640-6

RTECSNumber : AC8450000

TSCA : Yes

Recommended Storage : Ambient temperatures