Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfacetamide Sodium, Monohydrate, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6209-17-2
C8H11N2NaO4S
254.24
CNTXYOLKLFYMSC-UHFFFAOYSA-M
sodium N-[(4-aminophenyl)(oxido)oxo-λ⁶-sulfanylidene]acetamide hydrate
O.[Na+].CC(=O)N=S([O-])(=O)C1=CC=C(N)C=C1
6209-17-2
C8H11N2NaO4S
254.24
CNTXYOLKLFYMSC-UHFFFAOYSA-M
sodium N-[(4-aminophenyl)(oxido)oxo-λ⁶-sulfanylidene]acetamide hydrate
O.[Na+].CC(=O)N=S([O-])(=O)C1=CC=C(N)C=C1

SU124, 6209-17-2, C8H9N2NaO3S•H2O

Spectrum™ Sulfacetamide Sodium, Monohydrate, USP is an antibiotic that is used to treat skin infections and urinary tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Water 0.081
Quantity 5 kg
pH 8.0 to 9.5
CAS Max % 1
Percent Purity 99 to 100.5%
Grade USP