Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfacetamide sodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog Number AAB2242622
Click to view available options
Quantity:
25 g
100 g
500 g
6209-17-2
C8H9N2NaO3S
236.22
MFCD00007885
PQMSFAORUFMASU-UHFFFAOYSA-M
sodium sulfacetamide, sulfacetamide sodium monohydrate, sulfacetamide sodium salt hydrate, sulfacetamide sodic hydrate, unii-4nrt660kjq, sulfacetamide sodium usp, cetamide, sulfair forte, sulfair 10, 4nrt660kjq
6419954
CHEBI:63858
sodium;acetyl-(4-aminophenyl)sulfonylazanide;hydrate
CC(=O)N([Na])S(=O)(=O)C1=CC=C(N)C=C1
6209-17-2
C8H9N2NaO3S
236.22
MFCD00007885
PQMSFAORUFMASU-UHFFFAOYSA-M
sodium sulfacetamide, sulfacetamide sodium monohydrate, sulfacetamide sodium salt hydrate, sulfacetamide sodic hydrate, unii-4nrt660kjq, sulfacetamide sodium usp, cetamide, sulfair forte, sulfair 10, 4nrt660kjq
6419954
CHEBI:63858
sodium;acetyl-(4-aminophenyl)sulfonylazanide;hydrate
CC(=O)N([Na])S(=O)(=O)C1=CC=C(N)C=C1

Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

Solubility
Freely soluble in water. Insoluble in chloroform, ether, and benzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 100 g
Solubility Information Freely soluble in water. Insoluble in chloroform,ether,and benzene.
Formula Weight 236.23 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Sulfacetamide sodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-848-4
RTECSNumber AC8480000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit