Alfa Aesar™ Sulfadiazine, 99%

Manufacturer: Alfa Aesar™ A1237030

Quantity 250g

 View more versions of this product

Catalog No. AAA1237030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68-35-9
    Molecular Formula C10H10N4O2S
    Formula Weight 250.28
    MDL Number MFCD00006065
    Merck Index 14,8903
    Synonym N(1)-(2-Pyrimidinyl)sulfanilamide
    Chemical Name or Material Sulfadiazine

    Specifications

    Formula Weight 250.28
    Assay Percent Range 99%
    Health Hazard 2 GHS H Statement
    H334-H302-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 68-35-9
    Synonym N(1)-(2-Pyrimidinyl)sulfanilamide
    Merck Index 14,8903
    Health Hazard 1 Danger
    RTECS Number WP1925000
    Quantity 250g
    Molecular Formula C10H10N4O2S
    MDL Number MFCD00006065
    Melting Point 253° to 256°C
    Chemical Name or Material Sulfadiazine
    Assay 99%

    Description

    Applications
    Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)

    Solubility
    Soluble in dimethyl sulfoxide.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H334-H302-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit