Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadiazine, Powder, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
68-35-9
C10H10N4O2S
250.28
SEEPANYCNGTZFQ-UHFFFAOYSA-N
4-amino-N-(pyrimidin-2-yl)benzene-1-sulfonamide
NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CC=N1
68-35-9
C10H10N4O2S
250.28
SEEPANYCNGTZFQ-UHFFFAOYSA-N
4-amino-N-(pyrimidin-2-yl)benzene-1-sulfonamide
NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CC=N1

SU126, 68-35-9, C10H10N4O2S

Spectrum™ Sulfadiazine, Powder, USP is an antibiotic used to treat urinary tract infections and burns. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Ignition Residue 0.001
Quantity 500 g
Loss on Drying 0.005
CAS Max % 1
Percent Purity 98 to 102%
Grade USP