Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfadiazine Sodium, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
547-32-0
C10H9N4NaO2S
272.26
JLDCNMJPBBKAHH-UHFFFAOYSA-N
sodium 4-{[(pyrimidin-2-yl)azanidyl]sulfonyl}aniline
[Na+].NC1=CC=C(C=C1)S(=O)(=O)[N-]C1=NC=CC=N1
547-32-0
C10H9N4NaO2S
272.26
JLDCNMJPBBKAHH-UHFFFAOYSA-N
sodium 4-{[(pyrimidin-2-yl)azanidyl]sulfonyl}aniline
[Na+].NC1=CC=C(C=C1)S(=O)(=O)[N-]C1=NC=CC=N1

S1611, 547-32-0, C10H9N4NaO2S

Spectrum™ Sulfadiazine Sodium, USP is an antibiotic used to treat urinary tract infections and burns. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Loss on Drying 0.005
Quantity 5 kg
CAS Max % 1
Percent Purity 99 to 100.5%
Grade USP