Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethizole, ≥99% (HPLC), MilliporeSigma™ Supelco™

Sulfamethizole is an antibacterial drug used in the treatment of the urinary tract infections.

$51.30 - $103.50

Specifications

Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Linear Formula C9H10N4O2S2
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-3126
View Documents
MilliporeSigma Supelco
S563210G
10 g
Each for $51.30
Add to cart
 
11-101-3125
View Documents
MilliporeSigma Supelco
S563225G
25 g
Each for $103.50
Add to cart
 
Description

Description

Environmental and Pharmaceutical (small molecule)
Specifications

Specifications

≥99% (HPLC)
Analytical Standard
C9H10N4O2S2
Neat
Safety and Handling

Safety and Handling

RTECSNumber : WP0875000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit