Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, ACS, 99.3-100.3%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5329-14-6
5329-14-6
Packaging Poly Bottle
Chloride 1e-05
Heavy Metals 1e-05
Insoluble Matter 0.0001
Residue after Ignition 0.0001
Quantity 100 g
CAS Max % 1
Percent Purity 99.3 to 100.3%
Grade ACS

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.