Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Poly Bottle
5329-14-6
H3NO3S
97.088
IIACRCGMVDHOTQ-UHFFFAOYSA-N
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O
5329-14-6
H3NO3S
97.088
IIACRCGMVDHOTQ-UHFFFAOYSA-N
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O

High quality chemicals for laboratory and specialized industrial use

Quantity 100g
Formula Weight 97.09
Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Sulfamic Acid