Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Reagents
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
Specifications
Specifications
Chemical Name or Material | Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent |
Color | White to Off-White |
CAS | 5329-14-6 |
Packaging | Fiber Drum |
Specific Gravity | 2.15 |
Quantity | 50 lb. |
Molecular Formula | NH2SO3H |
Physical Form | Solid |
Synonym | Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid |
Molecular Weight (g/mol) | 97.0937 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction