Learn More
Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Reagents
Specifications
Specifications
| Chemical Name or Material | Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent |
| Color | White to Off-White |
| CAS | 5329-14-6 |
| Packaging | Poly Bottle |
| Specific Gravity | 2.15 |
| Quantity | 500 g |
| Molecular Formula | NH2SO3H |
| Physical Form | Solid |
| Synonym | Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid |
| Molecular Weight (g/mol) | 97.0937 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.