Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Reagents

Catalog No. 01337568
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-568 2.5 kg
01-337-569 10 kg
01-337-567 500 g
01-337-566 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-568 Supplier Reagents Holdings Llc Supplier No. C21143332.5A5
Only null left
Add to Cart
Add to Cart

Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sulfamic Acid, Technical Contains 0.3% Anti-caking Agent
Color White to Off-White
CAS 5329-14-6
Packaging Poly Bottle
Specific Gravity 2.15
Quantity 2.5 kg
Molecular Formula NH2SO3H
Physical Form Solid
Synonym Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid
Molecular Weight (g/mol) 97.0937
Grade Technical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.