Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilamide, BAKER™, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

63-74-1
C6H8N2O2S
172.202
FDDDEECHVMSUSB-UHFFFAOYSA-N
5333
CHEBI:45373

A grade of chemicals for laboratory use

Quantity 100g
CAS 63-74-1
Molecular Formula C6H8N2O2S
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Molecular Weight (g/mol) 172.202
PubChem CID 5333
Packaging Wide Mouth Amber Glass Bottle
ChEBI CHEBI:45373
Formula Weight 172.21
Specific Gravity 1L = 1.08kg
Chemical Name or Material Sulfanilamide
Show More Show Less