Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
121-57-3
C6H7NO3S
173.186
MFCD00007886
HVBSAKJJOYLTQU-UHFFFAOYSA-N
sulfanilic acid,sulphanilic acid,p-aminobenzenesulfonic acid,sulfanilsaeure,aniline-4-sulfonic acid,benzenesulfonic acid, 4-amino,aniline-p-sulfonic acid,aniline-p-sulphonic acid,p-aminophenylsulfonic acid,sulfanilicacid
8479
CHEBI:27500
4-aminobenzenesulfonic acid
C1=CC(=CC=C1N)S(=O)(=O)O
121-57-3
C6H7NO3S
173.186
MFCD00007886
HVBSAKJJOYLTQU-UHFFFAOYSA-N
sulfanilic acid,sulphanilic acid,p-aminobenzenesulfonic acid,sulfanilsaeure,aniline-4-sulfonic acid,benzenesulfonic acid, 4-amino,aniline-p-sulfonic acid,aniline-p-sulphonic acid,p-aminophenylsulfonic acid,sulfanilicacid
8479
CHEBI:27500
4-aminobenzenesulfonic acid
C1=CC(=CC=C1N)S(=O)(=O)O
Color Undesignated
Infrared Spectrum Authentic
Water 0.2% max. (K.F.)
Packaging Plastic bottle
Quantity 1kg
Assay Percent Range 99%
Linear Formula (H2N)C6H4SO3H
Formula Weight 173.19
Physical Form Liquid
Percent Purity ≥98.5% (Acid)
Chemical Name or Material Sulfanilic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.

GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning