Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfapyridine, Powder, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
144-83-2
C11H11N3O2S
249.29
GECHUMIMRBOMGK-UHFFFAOYSA-N
4-amino-N-(pyridin-2-yl)benzene-1-sulfonamide
NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
144-83-2
C11H11N3O2S
249.29
GECHUMIMRBOMGK-UHFFFAOYSA-N
4-amino-N-(pyridin-2-yl)benzene-1-sulfonamide
NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1

SU138, 144-83-2, C11H11N3O2S

Spectrum™ Sulfapyridine, Powder, USP is a sulfonamide antibacterial. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Melting Range 190-193°C
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 500 g
CAS Max % 1
Percent Purity 99 to 100.5%
Grade USP