Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfasalazine, USP, 97-101.5%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
599-79-1
C18H14N4O5S
398.39
OQANPHBRHBJGNZ-BKUYFWCQSA-N
(3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
599-79-1
C18H14N4O5S
398.39
OQANPHBRHBJGNZ-BKUYFWCQSA-N
(3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1

S1668, 599-79-1, C18H14N4O5S

Spectrum™ Chemical Sulfasalazine, USP belongs to a class of drugs called sulfa drugs and is used in the treatment of rheumatoid arthritis and some other autoimmune conditions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Ignition Residue 0.005
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 97 to 101.5%
Grade USP