Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfatase-2/SULF2 Mouse anti-Human, Mouse, Allophycocyanin/Cy7, Clone: 2B4, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP136727APCCY7
Description
Sulfatase-2/SULF2 Monoclonal antibody specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Sulfatase-2/SULF2 | |
2B4 | |
Flow Cytometry | |
Conjugated, Purified | |
SULF2 | |
Partial recombinant human Sulfatase 2 isoform b [Accession No. NP_940998]. | |
Protein G purified | |
Lipid and Metabolism | |
Primary | |
55959.0 | |
Human, Mouse |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Does not recognize human or mouse Sulfatase 1 or other family members. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title