Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfathiazole, Powder, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
10 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
72-14-0
C9H9N3O2S2
255.31
JNMRHUJNCSQMMB-UHFFFAOYSA-N
4-amino-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide
NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1
72-14-0
C9H9N3O2S2
255.31
JNMRHUJNCSQMMB-UHFFFAOYSA-N
4-amino-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide
NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1

S1258, 72-14-0, C9H9N3O2S2

Spectrum™ Sulfathiazole, Powder, USP is a short-acting sulfa drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 10 kg
Melting Range 200-204°C
Acidity To pass test
CAS Max % 1
Percent Purity 99.0 to 100.5%
Grade USP