Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfisoxazole, Crystalline, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

SU145, 127-69-5, C11H13N3O3S

Spectrum™ Sulfisoxazole, Crystalline, USP is a sulfonamide ("sulfa") antibiotic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Quantity 25 kg
Loss on Drying 0.005
Ignition Residue 0.001
Melting Range 194-199°C
CAS 127-69-5
CAS Max % 1
Molecular Formula C11H13N3O3S
InChI Key NHUHCSRWZMLRLA-UHFFFAOYSA-N
SMILES CC1=NOC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1C
IUPAC Name 4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzene-1-sulfonamide
Molecular Weight (g/mol) 267.30
Percent Purity 99 to 101%
Grade USP
Show More Show Less