Learn More
Bachem Sulfo-N-succinimidyl 4-maleimidobutyrate Sodium salt salt, 50mg, 185332-92-7, MFCD17215919 DFS Item
Manufacturer:  Bachem 4077463.0050
Sulfo-N-succinimidyl 4-maleimidobutyrate (Sulfo-GMBS) is a water-soluble, heterobifunctional crosslinking reagent for conjugation of cysteine-containing peptides to carrier proteins or surfaces.
Catalog No. 50-194-7396