Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfo Orange Solution, Fisher Chemical

Indicator for pH range 11.0-12.6

Catalog No. SS433-500



Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications

Specifications

Sulfo Orange Solution
0.05% Max.
Liquid
Pass Test
Yellow to Orange
547-57-9, 64-02-8, 1310-73-2, 7732-18-5, 5064-31-3
MFCD00007499
Glass Bottle
1.0g/cm³
500mL
Safety and Handling

Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation.
NFPA
Health: 0
Flammability:

Signal Word: CAUTION!

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit