Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfobromophthalein sodium hydrate, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
123359-42-2
C20H8Br4Na2O10S2·xH2O
837.76
MFCD16556312
HXUITPQMHVLBNV-LIYVDSPJSA-L
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
16211419
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)S(=O)(=O)[O-])C3=C(C(=C(C(=C3Br)Br)Br)Br)C(=O)O)S(=O)(=O)[O-])O.[Na+].[Na+]
123359-42-2
C20H8Br4Na2O10S2·xH2O
837.76
MFCD16556312
HXUITPQMHVLBNV-LIYVDSPJSA-L
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
16211419
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate
C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)S(=O)(=O)[O-])C3=C(C(=C(C(=C3Br)Br)Br)Br)C(=O)O)S(=O)(=O)[O-])O.[Na+].[Na+]
Water 8% max. (K.F.)
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color White
Solubility Information Solubility in water: soluble
Formula Weight 837.76
Physical Form Crystalline Powder
Percent Purity ≥95%
Chemical Name or Material Sulfobromophthalein sodium hydrate

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protecti

GHS Signal Word: Danger