Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfobromophthalein sodium hydrate, Thermo Scientific Chemicals

Catalog Number AC173401000
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
123359-42-2
C20H8Br4Na2O10S2
837.99
MFCD16556312
GHAFORRTMVIXHS-UHFFFAOYSA-L
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
16211419
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate
[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O
123359-42-2
C20H8Br4Na2O10S2
837.99
MFCD16556312
GHAFORRTMVIXHS-UHFFFAOYSA-L
disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
16211419
disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate
[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 100 g
Solubility Information Solubility in water: soluble
Formula Weight 837.76
Percent Purity ≥95%
Physical Form Crystalline Powder
Chemical Name or Material Sulfobromophthalein sodium hydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protecti

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit