Alfa Aesar™ Sulfobromophthalein sodium salt hydrate

Manufacturer: Alfa Aesar™ 4387803

Quantity 1g

 View more versions of this product

Catalog No. AA4387803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71-67-0
    Molecular Formula C20H8Br4O10S2Na2·xH2O
    Formula Weight 837.98 (Anhydrous)
    MDL Number MFCD00150017
    Merck Index 14,8954
    Synonym Bromosulfalein
    Chemical Name or Material Sulfobromophthalein sodium salt hydrate

    Specifications

    Formula Weight 837.98 (Anhydrous)
    Physical Form Powder
    MDL Number MFCD00150017
    CAS 71-67-0
    Merck Index 14,8954
    Synonym Bromosulfalein
    RTECS Number SM8395000
    TSCA TSCA
    Molecular Formula C20H8Br4O10S2Na2·xH2O
    Color White
    Sensitivity Moisture Sensitive
    Quantity 1g
    Chemical Name or Material Sulfobromophthalein sodium salt hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit