Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sulfolane, 80% w/w aq. soln.

Manufacturer:  Alfa Aesar™ H32505AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH32505AP


  Check Availability
Add to cart

Specifications

Description

Applications
Sulfolane is used in the chemical industry as a solvent for extractive distillation and chemical reactions. It is used to separate aromatic hydrocarbons (benzene, toluene and xylenes), to separate n-propyl alcohol and sec-butyl alcohol. It is also used to purify natural gas streams and for fractionation of fatty acids into saturated and unsaturated components and as a reaction solvent for the preparation of aromatic sulfonic acids, pyridines, isocyanates and pharmaceuticals. It can also be involved in the halogen exchange process and polymerization process. It is used as a plasticizer and curing agent.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

126-33-0
120.17
107765
Tetramethylene sulfone; Tetrahydrothiophene 1,1-dioxide
C4H8O2S
MFCD00005484
14,8955
Sulfolane

Specifications

80% w/w aq. soln.
126-33-0
120.17
107765
Tetramethylene sulfone; Tetrahydrothiophene 1,1-dioxide
Weak, Characteristic
500mL
C4H8O2S
MFCD00005484
14,8955
0° to 5°C
Sulfolane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P262-P501a
Do not get in eyes, on skin, or on clothing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XN0700000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit