Learn More
Aat BioquestSupplier Diversity Partner Sulforhodamine 101 *CAS 60311-02-6* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 76
Sulforhodamine 101 is a fluorescence reference standard for measuring fluorescence quantum yield. It is also the key precursor used for preparing rhodamine 101 sulfonyl chloride (also called Texas Red®, a trademark of Molecular Probes).
Catalog No. 50-195-2700