Learn More
Aat BioquestSupplier Diversity Partner Sulforhodamine B *CAS 3520-42-1* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 72
Sulforhodamine B, also known as Kiton Red, is primarily used as a polar tracer. It does not exhibit pH-dependent absorption or fluorescence over the range of 3 to 10.
Catalog No. 50-195-2696