Learn More
Aat BioquestSupplier Diversity Partner Sulforhodamine G *CAS 5873-16-5* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 73
Sulforhodamine G is used as a polar tracer as sulforhodamine B. Compared to rhodamine B it is more fluorescent. It does not exhibit pH-dependent absorption or fluorescence over the range of 3 to 10.
Catalog No. 50-195-2697