Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfosalicylic Acid, 3% (w/v) Aqueous Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
5965-83-3,7732-18-5
5965-83-3,7732-18-5
Percent Purity 3% ± 0.3%
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.