Learn More
Sulfotyrosine Mouse anti-Non-species specific, DyLight 650, Clone: 7C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259384C
Catalog No. NBP259384C
Add to cart
Description
Description
Sulfotyrosine Monoclonal antibody specifically detects Sulfotyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Sulfotyrosine
7C5
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine.