Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfotyrosine Antibody (7C5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259384F
Description
Sulfotyrosine Monoclonal specifically detects Sulfotyrosine in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Sulfotyrosine | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
100 μL | |
Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine. | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
7C5 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction