Learn More
Sulfotyrosine Mouse anti-Non-species specific, HRP, Clone: 7C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259384H
Catalog No. NBP259384H
Add to cart
Description
Description
Sulfotyrosine Monoclonal antibody specifically detects Sulfotyrosine in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Sulfotyrosine
7C5
Western Blot, ELISA
Purified
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA
HRP
PBS with No Preservative
Mouse
IgG1
100 ul
Primary
Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine.