Alfa Aesar™ Sulfur in Diesel Fuel standard solution, Specpure™, 400µg/g (0.040%)

Manufacturer: Alfa Aesar™ 38768AE

Quantity 100mL

 View more versions of this product

Catalog No. AA38768AE

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD01095233
    Chemical Name or Material Sulfur in Diesel Fuel standard solution, 400μg/g (0.040%)
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H351-H315-H319-H335-H227
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Quantity 100mL
    TSCA TSCA
    MDL Number MFCD01095233
    Health Hazard 1 Warning
    Chemical Name or Material Sulfur in Diesel Fuel standard solution, 400μg/g (0.040%)
    Grade Specpure™

    GHS H Statement
    H351-H315-H319-H335-H227
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit