Alfa Aesar™ Sulfur in Isooctane standard solution, Specpure™, 3000µg/g (0.300%)

Manufacturer: Alfa Aesar™ 41325AE

Quantity 100mL

 View more versions of this product

Catalog No. AA41325AE

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00804269
    UN Number UN1262
    Chemical Name or Material Sulfur in Isooctane standard solution, 3000μg/g (0.300%)
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 100mL
    TSCA TSCA
    UN Number UN1262
    Health Hazard 1 Danger
    MDL Number MFCD00804269
    Health Hazard 2 GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
    Chemical Name or Material Sulfur in Isooctane standard solution, 3000μg/g (0.300%)
    Grade Specpure™

    GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit