Learn More
Sulfur, plasma standard solution, S 1000µg/mL, Alfa Aesar™, Specpure™
Manufacturer:  Alfa Aesar 13842AD
Quantity 50mL
 View more versions of this product
Catalog No. AA13842AD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
Liquid
(NH4)2SO4 in H2O
MFCD00085316
Sulfur, plasma standard solution
Specifications
Liquid
(NH4)2SO4 in H2O
Ammonium sulfate: 0.413%; Water: 99.587%
50mL
Liquid
MFCD00085316
23 hPa (17mm Hg) at 20°C
Sulfur, plasma standard solution