Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfur, Precipitated (Powder/USP), Fisher Chemical
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Specifications
Boiling Point | 445°C |
Melting Point | 113°C |
Color | Yellow |
Physical Form | Solid |
Quantity | 500 g |
Assay Percent Range | 99.5 to 100.5 % |
Formula Weight | 32.07 |
Percent Purity | 99.5 to 100.5% |
Grade | USP |
Identification | Pass Test |
Show More |
Safety and Handling
Hazard Category | Signal Word: DANGER! |
Precautionary Statement | Emergency Overview Flammable solid. Causes eye and skin irritation and possible burns. Irritating to respiratory system. NFPA Health: |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction