Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur trioxide-pyridine complex, 98%, active SO3 ca 48-50%, Thermo Scientific Chemicals

Catalog Number p-7081981
Click to view available options
Quantity:
10 g
50 g
250 g
1000 g
26412-87-3
C5H5NO3S
159.16
MFCD00012437
UDYFLDICVHJSOY-UHFFFAOYSA-N
pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
168533
O=S(=O)=O.C1=CC=NC=C1
26412-87-3
C5H5NO3S
159.16
MFCD00012437
UDYFLDICVHJSOY-UHFFFAOYSA-N
pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
168533
O=S(=O)=O.C1=CC=NC=C1

Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfur trioxide-pyridine complex is used for sulfations of alcohols, sulfonations, deoxygenations and other reductions, oxidations with dimethyl sulfoxide. It is also used as a source of sulfur trioxide especially in the preparation of sulfate esters. Further, it is used as an activating electrophile in a Parikh-Doering oxidation. In addition to this, it is also used in the preparation and anticoagulation test of sulfated silk fibroin.

Solubility
Soluble in polar organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼155°C to 165°C
Quantity 10 g
UN Number UN3261
Beilstein 3704116
Sensitivity Moisture sensitive
Solubility Information Soluble in polar organic solvents.
IUPAC Name pyridine;sulfur trioxide
Formula Weight 159.16
Percent Purity 98%
Chemical Name or Material Sulfur trioxide-pyridine complex, Active SO3 ≈48 to 50%
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 247-683-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit