Sulfuric acid, 0.05N Standardized Solution, Alfa Aesar™

Manufacturer: Alfa Aesar 45048K2

Quantity 1L

 View more versions of this product

Catalog No. AA45048K2

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula H2SO4
    Formula Weight 98.08
    MDL Number MFCD00064589
    Chemical Name or Material Sulfuric acid

    Specifications

    Formula Weight 98.08
    Quantity 1L
    Color Colorless
    Physical Form Liquid
    Molecular Formula H2SO4
    MDL Number MFCD00064589
    Appearance Colourless solution
    Chemical Name or Material Sulfuric acid

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit