Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.1142 Normal, Suitable for Kjeldahl Nitrogen Analysis of Feed Grains, Ricca Chemical

Click to view available options
Quantity:
4L
20L
Packaging:
Cubitainer
Poly natural
7664-93-9, 7732-18-5
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
7664-93-9, 7732-18-5
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O

For Analysis of Suitable For Kjeldahl Nitrogen Analysis of Feed Grains

Concentration or Composition Notes 0.1142Normal
Color Colorless
Quantity 4L
CAS Min % 97.0, 0.57
CAS Max % 100.0, 0.6
Packaging Poly natural
Physical Form Liquid
Chemical Name or Material Sulfuric Acid