Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, 0.1N (0.05M) standard solution, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Plastic Bottle
Plastic jerrycan
7664-93-9,7732-18-5
H2O4S
98.072
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
7664-93-9,7732-18-5
H2O4S
98.072
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
Volumetric standard solution
Color Colorless
Quantity 2.5L
Additional Information Vapor Pressure: 1mmHg at 20°C
Linear Formula H2SO4
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15, 9104
Concentration 0.0998 to 0.1002N (20°C)
Density 1.3g/mL
Formula Weight 98.07
Physical Form Liquid
Grade Pure
Packaging Plastic Bottle
Solubility Solubility in water: miscible.
Boiling Point 100.0°C
Chemical Name or Material Sulfuric acid, 0.1N (0.05M) Standard Solution
Show More Show Less

GHS H Statement:
May be corrosive to metals.

GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning