Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.200 N, Reagents

Catalog No. 01336352
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Catalog No. Quantity
01-336-352 500 mL
01-336-356 10 L
01-336-355 1 L
01-336-354 20 L
01-336-353 4 L
5 options

Catalog No. 01-336-352

Supplier: Reagents Holdings Llc C7305520500A

Only null left
Add to Cart
Add to Cart

Sulfuric Acid, 0.200 N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH 1
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0067
Quantity 500 mL
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 0.200 N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.