Sulfuric acid, 0.5N Standardized Solution, Alfa Aesar™

Manufacturer: Alfa Aesar 35653-K2

Quantity 1L

 View more versions of this product

Catalog No. AA35653K2

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7664-93-9,7732-18-5
    Physical Form Liquid
    Molecular Formula H2SO4
    Formula Weight 98.08g/mol
    MDL Number MFCD00064589
    Chemical Name or Material Sulfuric acid

    Specifications

    Appearance Colorless to pale yellow liquid
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Color Colorless to pale yellow
    Quantity 1L
    CAS 7664-93-9,7732-18-5
    Physical Form Liquid
    Molecular Formula H2SO4
    Formula Weight 98.08g/mol
    MDL Number MFCD00064589
    Concentration or Composition (by Analyte or Components) Sulfuric acid: 5%; Water: 95%
    Health Hazard 1 H315-H319: Causes skin irritation. Causes serious eye irritation.
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 231-639-5
    Chemical Name or Material Sulfuric acid

    H315-H319: Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit