Sulfuric Acid, 1.000 +/- 0.001 N, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner R830000012X1A

Packaging Poly natural
Quantity 12 x 1L

 View more versions of this product

Catalog No. 83-003-2CS

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7732-18-5,7664-93-9
    Physical Form Liquid
    Assay Percent Range 1.000 ± 0.001 N at 20°C
    Chemical Name or Material Sulfuric Acid, 1.00 Normal

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Quantity 12 x 1L
    Solvent or Matrix Aqueous Solution
    Assay Percent Range 1.000 ± 0.001 N at 20°C
    CAS 7732-18-5,7664-93-9
    Packaging Poly natural
    Chemical Name or Material Sulfuric Acid, 1.00 Normal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit