Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, 15% fuming, 12-17% free SO3, Alfa Aesar™

Manufacturer:  Alfa Aesar 4554222

Quantity 100g

 View more versions of this product

Catalog No. AA4554222

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

8014-95-7
15% fuming, 12-17% free SO3
MFCD00137792
Sulfuric acid; 15% fuming
Liquid
H2SO4 + SO3
Oleum

Specifications

100g
Liquid
H2SO4 + SO3
Oleum
8014-95-7
15% fuming, 12-17% free SO3
MFCD00137792
Sulfuric acid; 15% fuming
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit