Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 18.0 N, Ricca Chemical
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Natural Poly Bottle
Safety-coated Glass Bottle
Specifications
Color | Colorless |
Appearance of Solution | Colorless Liquid |
Concentration | 17.95 to 18.05 N |
CAS Min % | 51.0, 47.0 |
CAS Max % | 53.0, 48.92 |
Packaging | Natural Poly Bottle |
Quantity | 500 mL |
Physical Form | Liquid |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
Assay | Normal at 20°C |
Show More |
Safety and Handling
ShelfLife | 36 months |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction