Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 2.0 N Solution, Spectrum™

Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle

S-640, 7664-93-9

Spectrum™ Sulfuric Acid, 2.0 N Solution - Spectrum utilizes the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Quantity 1 L
CAS 7664-93-9,7732-18-5
CAS Max % 0.1,0.9
Molecular Formula H2O4S
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Concentration 2 N +/-0.04
Molecular Weight (g/mol) 98.07
Grade Chemical Solution
Packaging Poly Bottle
Show More Show Less