Sulfuric acid, 20% fuming, 18-24% free SO3, Alfa Aesar™

Manufacturer: Alfa Aesar 4554322

Quantity 100g

 View more versions of this product

Catalog No. AA4554322

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 8014-95-7
    Physical Form Liquid
    Assay Percent Range 20% fuming, 18-24% free SO3
    Molecular Formula H2SO4 + SO3
    MDL Number MFCD00137792
    Synonym Oleum
    Chemical Name or Material Sulfuric acid; 15% fuming

    Specifications

    Quantity 100g
    Physical Form Liquid
    Density 1.9
    Assay Percent Range 20% fuming, 18-24% free SO3
    Molecular Formula H2SO4 + SO3
    MDL Number MFCD00137792
    CAS 8014-95-7
    Synonym Oleum
    Chemical Name or Material Sulfuric acid; 15% fuming

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit