Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 20% Fuming, ACS Analytical Reagent), Macron Fine Chemicals™

Catalog No. MK2886500
Click to view available options
Quantity:
500 g

Chemical Identifiers

CAS 7446-11-9
Molecular Formula O3S
Molecular Weight (g/mol) 80.06
InChI Key AKEJUJNQAAGONA-UHFFFAOYSA-N
Synonym sulphur trioxide, sulfite radical anion, sulfur trioxide, contains stabilizer, sulfan, oxosulfane dioxide, sulfonylideneoxidane, unii-hh2o7v4lyd, hh2o7v4lyd, so3
PubChem CID 24682
ChEBI CHEBI:29384
IUPAC Name sulfur trioxide
SMILES O=S(=O)=O

Specifications

Density 1.91kg/L
Quantity 500 g
Formula Weight 178.14
Grade ACS
Chemical Name or Material Sulfuric Acid, 20%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.